Corning® Collection Containers, Corning | Avantor
Corning® Collection Containers, Corning
Supplier:   Corning
Corning® Collection Containers, Corning
Supplier:  Corning
Certificates
Corning® Collection Containers, Corning
15 Options Available Below
Product Details & Documents
Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.
  • Single-use bags
  • Designed to be practical and cost-effective alternatives to rigid-walled containers
  • Ideal for biopharmaceutical applications
Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.
Product Family Options
Informations sur le produit
  • Capacité
    Matière
    Port 1
    Port 2
    Port 3
    Availability
    Price
  • 24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Female Luer with ¹/₄" Barb, cap, Slide clamp"
    24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Male Luer with ¹/₄" Barb, cap, Slide clamp"
    Discontinued
    Supplier Number: -
  • 24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Female Luer with ¹/₄" Barb, cap, Slide clamp"
    24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Male Luer with ¹/₄" Barb, cap, Slide clamp"
    Discontinued
    Supplier Number: -
  • 24 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Male CPC connector with ³/₈" Barb, PVC dust cover cap, pinch clamp"
    24 L × ³/₈ Int.Ø ×⁵/₈" Ext.Ø tubing, Male CPC connector with ³/₈" Barb, PVC dust cover cap, pinch clamp"
    36 L × ¹/₈ Int.Ø ×¹/₄" Ext.Ø tubing, Female Luer with ¹/₈" Barb, Male Luer cap"
    Discontinued
    Supplier Number: -
  • 1 L
    EVA
    Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp
    Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp
    Female Luer with ³⁄₁₆ barb, swabbable valve, cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1l)
    Each (1l) - $87.90
    $87.90
    Each (1l)
       $87.90
    Supplier Number: 91-200-01
  • 2 L
    EVA
    Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp
    Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp
    Female Luer with ³⁄₁₆ barb, swabbable valve, cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (2l)
    Each (2l) - $101.00
    $101.00
    Each (2l)
       $101.00
    Supplier Number: 91-200-02
  • 5 L
    EVA
    Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp
    Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp
    Female Luer with ³⁄₁₆ barb, swabbable valve, cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (5l)
    Each (5l) - $116.00
    $116.00
    Each (5l)
       $116.00
    Supplier Number: 91-200-05
  • 10 L
    EVA
    Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp
    Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp
    Female Luer with ³⁄₁₆ barb, swabbable valve, cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (10l)
    Each (10l) - $127.00
    $127.00
    Each (10l)
       $127.00
    Supplier Number: 91-200-10
  • 20 L
    EVA
    Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp
    Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp
    Female Luer with ³⁄₁₆ barb, swabbable valve, cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (20l)
    Each (20l) - $131.00
    $131.00
    Each (20l)
       $131.00
    Supplier Number: 91-200-20
  • 10 L
    EVA
    Female MPC connector with ¼ barb + plug, slide clamp
    Male MPC connector with ¼ barb + plug, slide clamp
    Vinyl cap, swabbable valve, female Luer with barb
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (10l)
    Each (10l) - $99.10
    $99.10
    Each (10l)
       $99.10
    Supplier Number: 91-200-36
  • 20 L
    EVA
    Female MPC connector with ¼ barb + plug, slide clamp
    Male MPC connector with ¼ barb + plug, slide clamp
    Vinyl cap, swabbable valve, female Luer with barb
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (20l)
    Each (20l) - $130.00
    $130.00
    Each (20l)
       $130.00
    Supplier Number: 91-200-39
  • 500 ml
    EVA
    Break-off Injection site
    Female MPC connector with ¼ barb, male MPC plug
    Male MPC connector with ¼ barb, female MPC cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500ml)
    Each (500ml) - $97.00
    $97.00
    Each (500ml)
       $97.00
    Supplier Number: 91-200-41
  • 1 L
    EVA
    Break-off Injection site
    Female MPC connector with ¼ barb, male MPC plug
    Male MPC connector with ¼ barb, female MPC cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1l)
    Each (1l) - $79.80
    $79.80
    Each (1l)
       $79.80
    Supplier Number: 91-200-42
  • 5 L
    EVA
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, male MPC connector with ⅜" barb, female MPC cap
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, female MPC connector with ⅜" barb, male MPC cap
    Break-off Injection site
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (5l)
    Each (5l) - $254.00
    $254.00
    Each (5l)
       $254.00
    Supplier Number: 91-200-43
  • 10 L
    EVA
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, male MPC connector with ⅜" barb, female MPC cap
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, female MPC connector with ⅜" barb, male MPC cap
    Break-off Injection site
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (10l)
    Each (10l) - $188.00
    $188.00
    Each (10l)
       $188.00
    Supplier Number: 91-200-45
  • 20 L
    EVA
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, male MPC connector with ⅜" barb, female MPC cap
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, female MPC connector with ⅜" barb, male MPC cap
    Break-off Injection site
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (20l)
    Each (20l) - $204.00
    $204.00
    Each (20l)
       $204.00
    Supplier Number: 91-200-47
  • 50 L
    EVA
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, male MPC connector with ⅜" barb, female MPC cap
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, female MPC connector with ⅜" barb, male MPC cap
    Break-off Injection site
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (50l)
    Each (50l) - $228.00
    $228.00
    Each (50l)
       $228.00
    Supplier Number: 91-200-48
  • 100 L
    ULDPE
    60 L × ½" int.Ø × ¾" ext.Ø tubing, male MPX connector, female MPC cap, pinch clamp
    40 L × ⅛" int.Ø × ¼" ext.Ø tubing, female Luer with ⁵⁄₃₂" barb, swabbable valve, non-breather female Luer cap
    60 L × ¹/₂" int.Ø × ³/₄" ext.Ø tubing, 1.5" tri-clover with ¹/₂" barb, pinch clamp
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100l)
    Each (100l) - $369.00
    $369.00
    Each (100l)
       $369.00
    Supplier Number: 91-200-82
  • 200 L
    ULDPE
    60 L × ½" int.Ø × ¾" ext.Ø tubing, male MPX connector, female MPC cap, pinch clamp
    40 L × ⅛" int.Ø × ¼" ext.Ø tubing, female Luer with ⁵⁄₃₂" barb, swabbable valve, non-breather female Luer cap
    60 L × ¹/₂" int.Ø × ³/₄" ext.Ø tubing, 1.5" tri-clover with ¹/₂" barb, pinch clamp
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (200l)
    Each (200l) - $388.00
    $388.00
    Each (200l)
       $388.00
    Supplier Number: 91-200-83