HardyDisk™ AST , Hardy Diagnostics
HardyDisk™ AST’s are impregnated paper disks used for antimicrobial susceptibility testing (AST), also known as disk diffusion or Kirby-Bauer testing
Fully compatible with the BBL single or multiple disk dispensers.
: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Frequently Bought Together