SpeedStreaks™ Loops and Needles in Rigid Containers, Hardy Diagnostics
CA10753-956
- Color:Light Blue
- Description:Disposable Flexible (Soft) Inoculation Loop
- Size:10 μL
- Cat. no.:CA10753-956
- Packaging:Plastic
- Supplier No.:HST10F
SpeedStreaksTM are disposable inoculating loops, available in several styles and are color coded for convenience
Hexagonal loop shaft improves grip, while the ultra smooth loop surfaces allow for easy streaking. Loops are sterile and free of lubricants, oils, and electrostatic charges. Packaged in rectangular, plastic tube with lid that allows placement on clean table without rolling. 50 per tube, 1000 per box.
The 1μl and 10μl loops come each in two different styles, rigid, or flexible-soft, to fit a variety of streaking preferences. Certificate of Calibration included.