SpeedStreaks™ Loops and Needles, Calibrated, Hardy Diagnostics
Catalogue # CA10753-964
Fournisseur: HARDY DIAGNOSTICS CA
Spécifications
- Color:Violet
- Description:Rigid (Hard) Inoculating Needle
- Cat. no.:CA10753-964
- Supplier No.:HSND
Spécifications
À propos de cet article
SpeedStreaksTM are disposable inoculating loops, available in several styles and color coded for convenience.
The 1 μL and 10 μL loops come in two different styles (rigid or flexible) to fit a variety of streaking preferences.
Ordering information: Packaged in Ziploc® bags with certification of calibration.