ContactUsLinkComponent
MacConkey Agar (RSR), Hardy Diagnostics
Catalogue # CA10753-960
Fournisseur:  HARDY DIAGNOSTICS CA
Numéro CAS:  
undefined
MacConkey Agar (RSR), Hardy Diagnostics
Catalogue # CA10753-960
Fournisseur:  HARDY DIAGNOSTICS CA
Supplier Number:  GA35
Numéro CAS:  
Spécifications
  • Pack type:
    Plate
  • Description:
    MacConkey Agar
  • L×W:
    15 x 100 mm
  • Cat. no.:
    CA10753-960
  • Supplier No.:
    GA35
Spécifications
À propos de cet article
MacConkey Agar (RSR) is used as a selective and differential medium for the culture and isolation of Gram- negative bacteria, controls swarming of Proteus.
Recommended for use in the isolation of Gram- negative bacilli, including coliform organisms and enteric pathogens, on the basis of lactose fermentation.
Reduced stacking ring for automated inoculating devices.