To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Groundwater Pollution & Spill Assessment Lab Activity
- No. of Lab Groups:5
- Cat. no.:470023-786
- Supplier No.:WARD470023-786
Specifications
About this item
Innovative Introduction to Basic Groundwater Concepts and Problem-Solving.
- Construct topographic profiles and geologic cross sections
- Map the flow of a groundwater pollution plume
- Includes a teacher's guide
Give students real-world experience in environmental geology with this hands-on investigation. As working hydrogeologists, students are given a simulated groundwater pollution problem to assess possible contamination of area wells and direct a course of action for immediate remediation. They will construct a topographic profile and geological cross-section of the spill area, determine the porosity and permeability of subsurface strata, and test simulated water samples from the spill site to assess the relative concentration of the pollutant and map the direction of plume flow. You will receive materials for five setups, a teacher's guide, and student worksheets.