Corning® Low-Profile Universal Lid for 384-Well Microplates, with Corner Notch, Corning
Catalog # 89501-316
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Color:Clear
- Description:384-Well Low-Profile Universal Lid, with Corner Notch
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:89501-316
- Supplier No.:3908
Specifications
About this item
This universal lid with corner notch does not need to be oriented in any particular direction to be placed on Corning® microplates.
This lid also features a shorter skirt than standrard lids. Lid is manufactured from polystyrene.