Gusset and Flat Bags, Elkay Plastics
Fournisseur: Elkay Plastics
Certificats
À propos de cet article
Gusseted bags feature creased sides that expand to accommodate bulk items.
- Elkay polyethylene bags are made from virgin materials
Low density polyethylene (LDPE) bags are the most commonly used plastic bag because they provide protection for everyday use.
Certifications: Comply with FDA requirements for use in food applications.
Frequently Bought Together