Lab Loc® Specimen Bags, Elkay Plastics
Elkay’s Lab-Loc® reclosable seal top specimen bags with Seal-N-Rip™ removable biohazard symbols provide an means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs.
Printed in both English and Spanish and available in a variety of sizes, colors and printed messages for various types of specimens.
The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncominated. Their three wall construction provides a handy outer pouch to contain accompanying paperwork during specimen transfer.
: Bags are inner-packed in individually-marked and barcoded dispenser packs for convenience.
Frequently Bought Together