ContactUsLinkComponent
Surgical Skin Marker, Sklar
Surgical Skin Marker, Sklar
Catalog # 10163-480
Supplier:  Sklar
undefined
Surgical Skin Marker, Sklar
Catalog # 10163-480
Supplier:  Sklar
Supplier Number:  10-1440
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Surgical skin marker
  • Cat. no.:
    10163-480
  • Supplier No.:
    10-1440
Specifications
About this item
Sklar surgical skin markers are designed to allow surgeons to conveniently demarcate selected sites, prior to or during surgical procedures
Sterile. Scrub resistant ink. Durable tip. Non-smearing. Latex-free.