Disposable Inoculation Needle, Plastic
Catalogue # 30002-172
Fournisseur: Copan Diagnostics
Spécifications
- Color:Violet
- Description:Inoculating needle
- Cat. no.:30002-172
- Packaging:25/Ziplock Bag
- Supplier No.:COP-NED
Spécifications
À propos de cet article
COPAN Plastic Inoculation Needles have an ultra-smooth surface to avoid gouging agar
Plastic needles are free of lubricants, oils, and electrostatic charges.