Polypropylene Griffin Low-Form Beaker, 150 ml
150 ml (5 oz.), Polypropylene Griffin Low-Form Beakers
Catalog # 470211-734
Supplier: Thermo Fisher Scientific
Supplier #: 1201-0150
Polypropylene Griffin Low-Form Beaker, 150 ml
Catalog # 470211-734
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Carry out precise and efficient pouring with Polypropylene Griffin Low-Form Beakers, featuring a single-spout design and easy-to-read silk-screened graduations.
Specifications
- Capacité:150 ml (5 oz.)
- Désignation:Polypropylene Griffin Low-Form Beakers