PMP Erlenmeyer Flasks with Closure
125 ml
Catalog # 470205-834
Supplier: Thermo Fisher Scientific
Fournisseur #: 4109-0125
Jump to:
Product Details & Documents
Provide a Combination of Chemical Resistance, Strength and Transparency.
- Constructed with sturdy polymethylpentene
- Excellent of culture media preparation and storage
- Cap fits tightly with screw closure
Spécifications
- Capacité:125 ml