Supertek® Acceleration Timers
À propos de cet article
Calculate acceleration, drag, and friction by measuring the interval between dots.
- Mechanical timer device
- 60 Hz (mains power frequency)
- Timer size: 7L×2W×1¹/₄" H
- Tape length: 300 m per roll
- Tape width: 9 mm
- Carbon disk Ø: 38 mm
This timer works like a clock and produces precisely sixty dots per second on a moving strip of paper. The unit is heavy enough to rest on the edge of a table without clamps and is also designed to clamp to an upright support rod or crossbar. The timer is powered directly by 110 V AC via a 5' 10" cord with an on/off switch.
Ordering information: Timer is supplied with 12 carbon disks and instructions. Tape is required and sold seperately. Not available in Canada.
1 Options Available Below
Product Details & Documents
Calculate acceleration, drag, and friction by measuring the interval between dots.
- Mechanical timer device
- 60 Hz (mains power frequency)
- Timer size: 7L×2W×1¹/₄" H
- Tape length: 300 m per roll
- Tape width: 9 mm
- Carbon disk Ø: 38 mm
This timer works like a clock and produces precisely sixty dots per second on a moving strip of paper. The unit is heavy enough to rest on the edge of a table without clamps and is also designed to clamp to an upright support rod or crossbar. The timer is powered directly by 110 V AC via a 5' 10" cord with an on/off switch.
Ordering information: Timer is supplied with 12 carbon disks and instructions. Tape is required and sold seperately. Not available in Canada.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Acceleration timerDiscontinued-Supplier Number: PH10547
- Replacement carbon paper disks, pack of 24Each$37.25Pack of 24$36.90Case of 4800$6,721.85Supplier Number: P10563A
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement carbon paper disks, pack of 24