Mono-functional maleimides are particularly suitable for the selective labelling of molecules containing free sulfhydryl groups, such as cysteine residues in proteins and peptides and oligonucleotides.
- More selective method of antibody labelling that reduces the possibility of the label interfering with subsequent antibody-antigen reactions
- Provides an alternative when CyDye™ NHS esters are unsuitable
- Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings
They are also useful in antibody labelling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Storage conditions:2 - 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
- Form:Solid
- pH insensitive:3 pH Insensitive Min.; 10 pH Insensitive Max. (for 995017-553 only)
- Solubility:Soluble in cold water and hot water