Tuerk solution | Avantor
Tuerk solution
Tuerk solution
Certificates
Tuerk solution
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Turk's blood diluting fluid has a stain (gentian violet) plus 6% acetic acid and is ideal for manual leukocyte counting.
  • Ideal for Manual Leukocyte Counting
  • Six Percent Acetic Acid
  • Gentian Violet Stain
Stains the nuclei of the white blood cells making them easier to see and count.
Synonymes:  Turk solution, Acetic acid gentian violet solution
  • Numéro MDL:MFCD00148719
Documents
Product Family Options
Informations sur le produit
  • Taille
    Availability
    Price
  • 4 oz.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (4oz)
    Each (4oz) - $58.88
    $58.88
    Prix promotionnel pour les nouveaux clients
    Each (4oz)
       $58.88
    Supplier Number: ES930
  • 16 oz.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (16oz)
    Each (16oz) - $91.60
    $91.60
    Prix promotionnel pour les nouveaux clients
    Each (16oz)
       $91.60
    Supplier Number: ES931
  • 32 oz.
    Discontinued
    Supplier Number: ES932