ContactUsLinkComponent
Bent PEEK Cannula, Quality Lab Accessories
Catalogue # 89039-790
Fournisseur:  QLA
undefined
Bent PEEK Cannula, Quality Lab Accessories
Catalogue # 89039-790
Fournisseur:  QLA
Supplier Number:  CA8475-PV
Spécifications
  • Description:
    4.75" (900 mL) Bent PEEK Cannula
  • Size:
    12.07 cm (4.75")
  • Cat. no.:
    89039-790
  • Supplier No.:
    CAN475-PV