Right Angle Tee Joint to Tube Compression Adapter with Drip Tip, Ace Glass Incorporated
These glass standard taper joint adapters feature a drip tip and right angle tee ground glass tube sidearms.
- Glass [ST] joint
- Drip tip
- Right angle tee sidearms
The sidearms are used to make tube compression fitting connections to a wide variety of flexible and rigid tubing using PTFE fittings.
Frequently Bought Together