Digital Stirring Motor and Controller, Hazmat Series, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
A heavy duty, UL and CSA rated hazardous area motor with digital controller
The motor has variable speeds up to 1750 rpm with a maximum torque of 91.5". The motor output is 230 V 60 Hz, 3 phase. The motor shaft is 5/8". The Digital Microdrive control box has a compact wall mountable design, a six-digit LED display, flat panel control, and is IP66/NEMA 4X rated.
The stir motor meets Class I, Division I, Group D requirements for flammable liquids and gases, and Class II, Division II, Groups F&G for combustible dust.
Delivery: Available in 115 or 230V models, CE and cULus. The stirrer motor, controller, and gear reducer are sold together.
Frequently Bought Together