Individual Slotted Killogram Weights | Avantor
Individual Slotted Killogram Weights
Supplier:   Wards
Individual Slotted Killogram Weights
Supplier:  Wards
Certificates
Individual Slotted Killogram Weights
Individual Slotted Killogram Weights
3 Options Available Below
Product Details & Documents
Individual cast iron slotted weights.
  • Available individually
  • Stackable
  • Made from solid cast iron
  • Finished with a black enamel
  • Stamped with weight value
Slotted iron masses are suitable for use with sonometers or other applications, where a large load is required. The slotted masses can be hung on a hanger.
Ordering information: Hanger ordered separately.
Product Family Options
Informations sur le produit
  • Désignation
    Weight
    Availability
    Price
  • Slotted Weight
    1 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $40.90
    $40.90
    Each
       $40.90
    Supplier Number: PH0264B2
  • Slotted Weight
    2 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $75.45
    $75.45
    Each
       $75.45
    Supplier Number: PH0264B3
  • Slotted Weight
    5 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.05
    $136.05
    Each
       $136.05
    Supplier Number: PH0264B4
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Slotted Hanger Only, 0.5 kg
    Slotted Hanger Only, 0.5 kg
    470229-366
    Log in pour la disponibilité
    Each - $37.90