Cuvettes for Mindray Analyzers, Globe Scientific
Cuvettes for Mindray Analyzers, Globe Scientific
Supplier:   Globe Scientific
Cuvettes for Mindray Analyzers, Globe Scientific
Supplier:  Globe Scientific
Certificates
Cuvettes for Mindray Analyzers, Globe Scientific
Cuvettes for Mindray Analyzers, Globe Scientific
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These high quality consumables are designed for use with the Mindray® BS200 & BS300 chemistry analyzers.
Ordering information: Individually wrapped into four boxes of 250 (1000).
Product Family Options
Product Information
  • Désignation
    Matière
    Availability
    Price
  • Cuvette for BS200
    Polystyrene
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $1,502.63
    $1,502.63
    New Customer Promotional Price
    Case of 1,000
       $1,502.63
    Supplier Number: 5145
  • Cuvette for BS300
    Polystyrene
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $1,502.63
    $1,502.63
    New Customer Promotional Price
    Case of 1,000
       $1,502.63
    Supplier Number: 5146