About this item
Organic binder gives these filters higher wet strength and lower fiber release compared to binder-free grades.
Grade GC90 is recommended for fine particles and aerosols, clinical screening, or prefilter for membrane.
Grade DP70 has very high loading capacity. Recommended for liquids with high concentration of protein and other substances.
Grade GS25 is a thin filter with less loading capacity as others. Can be used for suspended solids analysis.
Recommended for high pressure filtration, applications requiring wet strength, or as a prefilter.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.