ContactUsLinkComponent
Resprep® Resin SPE Disks, Restek
  10015-396
 :  Restek
undefined
Resprep® Resin SPE Disks, Restek
  10015-396
 :  Restek
 :  26023
  • Description:
    Resprep® Resin SPE Disks, 47 mm I.D.
  • Cat. no.:
    10015-396
  • Supplier No.:
    26023
These 47 mm glass fiber disks are embedded with styrene/DVB resin.
Resprep® resin SPE disks are designed for use with chlorinated, benzidine-containing, or nitrogen-containing pesticides.
 : Disks meet requirements of EPA methods 515.2 and 553.