Resprep®-C18 and Resprep®-C8 SPE Disks, Restek
Fournisseur: Restek
Certificats
À propos de cet article
Glass fiber disks embedded with either C18 or C8 bonded silica are designed to extract semi-volatile organic compounds, and feature a deep-pore design that reduces clogging and allows faster flow rates.
- Designed for extraction of semi-volatile organic compounds
- Engineered to reduce clogging
These disks are more economical than traditional PTFE disks.
Certifications: Disks meet requirements for EPA methods 525.1, 506, 550.1, and 549.1.
Frequently Bought Together