ContactUsLinkComponent
Stable "Z"® Heated Lid and Controller, Bioptechs Inc.®
Stable "Z"® Heated Lid and Controller, Bioptechs Inc.®
Catalog # 10199-848
Supplier:  BIOPTECHS INC.
undefined
Stable "Z"® Heated Lid and Controller, Bioptechs Inc.®
Catalog # 10199-848
Supplier:  BIOPTECHS INC.
Supplier Number:  40319260812
Specifications
  • Description:
    Stable "Z"® Heated Lid and Controller
  • Cat. no.:
    10199-848
  • Supplier No.:
    40319260812
Specifications
About this item
Designed for use with the Stable "Z" system to prevent condensation and provide a more uniform temperature gradient across the dish of the Stable "Z".
  • Prevents condensation
  • Prevents outside contamination
Certifications: CE certified.
Delivery: Includes lid, controller, and power supply.