Interchangeable Coverslip Dish Sets, Bioptechs Inc.® | Avantor
Interchangeable Coverslip Dish Sets, Bioptechs Inc.®
Supplier:   BIOPTECHS INC.
Interchangeable Coverslip Dish Sets, Bioptechs Inc.®
Supplier:  BIOPTECHS INC.
Certificates
Interchangeable Coverslip Dish Sets, Bioptechs Inc.®
Interchangeable Coverslip Dish Sets, Bioptechs Inc.®
2 Options Available Below
Product Details & Documents
The Interchangeable Coverslip Dish Sets are autoclavable and reusable culture dishes
  • Separate coverglass enables the user to pre-treat or condition the coverslip if necessary
  • 25mm clear aperture for more translation room
  • Accessories available for fluid perfusion and 2nd optical surface
  • No metal components have contact with the specimen or media
  • Threaded tube can be made any height for special applications
  • All parts are autoclaveable
  • Significantly reduced Z axis drift as compared to plastic
  • Metal base transfers heat from peripheral warmers
  • 30mm German Desag glass ideal for cell culture
  • O-Ring sealed for positive leak prevention
  • Low cost coverslip replacement
  • Fits any 35mm dish warmer
  • Easy to assemble
Dishes are made of a stainless steel ring into which a 30mm, Desag 263 glass #1.5 coverslip is placed. The coverslip is retained by a leakproof, O-Ring sealed, 10m high polycarbonate threaded insert. Coverslip dish sets are available in two sizes, with either a 35mm or 40mm diameter.
Certifications: CE certified.
Ordering information: Sets include two O-Rings and fifty 30mm coverslips and ICD.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Interchangeable cover slip dish set, 40 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $363.68
    $363.68
    Kit of 1
       $363.68
    La quantité minimale de commande est de 1
    Supplier Number: 190310
  • Interchangeable cover slip dish sett, 35 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $657.15
    $657.15
    Kit of 1
       $657.15
    La quantité minimale de commande est de 1
    Supplier Number: 19031035