Glass Culture Cylinders and Cylinder Starter Set, Bioptechs Inc.®
An assortment of borosilicate glass cylinders, all measuring 5 mm high, each with an optically flat polished end to eliminate the need for grease when plating cells.
- Restrict and concentrate the growth of cells
- Heavier than plastic cloning rings
- Eliminate floating
- Seal without grease
- No contamination
This facilitates positioning for observation and improves plating time. Culture cylinders are reusable and autoclaveable.
: CE certified.
: Starter Set includes 2, 4, 6, 8, and 10 mm sizes.
Frequently Bought Together