Objective Thermal Isolators, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
Certificates
About this item
These 9 mm thick internally and externally threaded polymer rings are to be placed between an objective and nosepiece to inhibit thermal conductivity.
Available in three formats: for RMS, 0.8"×36 tpi, 25 mm for 25×0.75 mm objectives, or 27 mm for 27×0.75 mm objectives.
Certifications: CE certified.
Delivery: Each isolator includes an AR window to enable a dry air trap, thus preventing condensation on cooled objectives.
Frequently Bought Together