FCS2® Open Mode Adapter, Bioptechs Inc.®
Catalog # 10200-022
Supplier: BIOPTECHS INC.
Specifications
- Description:FCS2® Open Mode Adapter
- Cat. no.:10200-022
- Supplier No.:06031921513
Specifications
About this item
The white, top portion of a FCS2® without perfusion ports, which enables the FCS2® to be assembled without the microaqueduct slide
This enables sealing of the 40 mm coverslip to the base of the chamber. Oftentimes used for microinjection.
Certifications: CE certified.