Anti-IgG Donkey Polyclonal Antibody (CF™ 543)
Fournisseur: Biotium
Certificats
À propos de cet article
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 89411-076
- 89411-524
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 543
- CF® 543
- Reactivity:
- Sheep
- Sheep
- Host:
- Donkey
- Donkey
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Epitope:
- Heavy and Light chain
- Heavy and Light chain
- Cross adsorption:
- Minimal Cross-Reactivity: Ch, GP, Hs, Hu, Ms, Rb, Rt, SHm
- Minimal Cross-Reactivity: Ch, GP, Hs, Hu, Ms, Rb, Rt, SHm
Spécifications
Frequently Bought Together