Order Entry
Canada
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalogue # CA82022-756
Fournisseur:  G-Biosciences
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalogue # CA82022-756
Fournisseur:  G-Biosciences
Supplier Number:  786-R45
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Size:
    1 mL
  • Cat. no.:
    CA82022-756
  • Supplier No.:
    786-R45

Spécifications

À propos de cet article

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rat