Order Entry
Canada
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # CA82022-752
Supplier:  G-Biosciences
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # CA82022-752
Supplier:  G-Biosciences
Supplier Number:  786-R43
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Size:
    1 mL
  • Cat. no.:
    CA82022-752
  • Supplier No.:
    786-R43

Specifications

About this item

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse