Anti-IgG Goat Polyclonal Antibody (CF™ 790)
Catalog # 89427-198
Supplier: Biotium
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:CF® 790
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Size:50 µL
- Epitope:Heavy and Light chain
- Cross adsorption:Minimal Cross-Reactivity: Bv, Hs, Hu, Rb, Sw
- Cat. no.:89427-198
- Supplier No.:20342-50UL
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 790
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse