À propos de cet article
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™647 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™647 (Ex/Em 650/665 nm) is brighter than Cy®5 and an excellent alternative to Alexa Fluor® 647 for antibody conjugates.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 647
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10018-244
- 89138-470
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Mouse IgG
- Immunoglobulin G
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 647
- CF® 647
- Reactivity:
- Mouse
- Mouse
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Antibody fragment:
- F(ab')2
- F(ab')2
- Epitope:
- Heavy and Light Chain
- Heavy and Light chain
- Form:
- Liquid
- Liquid
- Purification:
- Affinity purified
- Affinity purified
- Concentration:
- 2 mg/mL
- 2 mg/mL
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide
- PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide
- Storage temperature:
- Store at –20 °C.
- Store at –20 °C.