Anti-IFN-g Mouse Monoclonal Antibody [clone: 4S.B3]
Supplier: Biolegend
Certificates
About this item
Purified anti-human IFN-γ [4S.B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: ELISA, WB, ICFC; Size: 500 μg
Each lot of this antibody is quality control tested by ELISA assay. For ELISA Capture applications, the antibody should be titrated between 0.25 - 2 μg/ml to determine optimal condition. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: 4S.B3
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus
Specifications
- Catalog No:
- CA502501-BL
- CA502502-BL
- Antigen symbol:
- IFN-g
- IFN-g
- Antigen synonyms:
- Type II interferon|T cell interferon|Macrophage-activating factor (MAF)|IFNG|Interferon-γ|Immune interferon
- Type II interferon|T cell interferon|Macrophage-activating factor (MAF)|IFNG|Interferon-γ|Immune interferon
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 4S.B3
- 4S.B3
- Reactivity:
- Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3458
- 3458
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Gel shift:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Partially purified, native human IFN-γ
- Partially purified, native human IFN-γ
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together