About this item
PerCP anti-human CD20 [2H7]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Baboon;Capuchin Monkey;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus;Squirrel Monkey
Specifications
- Catalog No:
- CA302323-BL
- CA302324-BL
- Antigen symbol:
- CD20
- CD20
- Antigen synonyms:
- Bp35|B1
- Bp35|B1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- Clone:
- 2H7
- 2H7
- Reactivity:
- Baboon, Capuchin monkey, Cynomolgus monkey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
- Baboon, Capuchin monkey, Cynomolgus monkey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
- Host:
- Mouse
- Mouse
- Gene ID:
- 931
- 931
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human tonsillar B cells
- Human tonsillar B cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PerCP under optimal conditions. The solution is free of unconjugated PerCP and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PerCP under optimal conditions. The solution is free of unconjugated PerCP and unconjugated antibody.
- Storage temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT