About this item
PE anti-human Perforin [dG9]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Bovine (Cow, Cattle); Apps: ICFC; Size: 100 tests
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: Perforin
Clonality: Monoclonal
Clone: dG9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Bovine (Cow;Cattle)
Specifications
- Catalog No:
- CA308105-BL
- CA308106-BL
- Antigen name:
- Perforin
- Perforin
- Antigen synonyms:
- lymphocyte pore-forming protein|cytolysin|lymphocyte pore forming protein|perforin-1|PRF1
- lymphocyte pore-forming protein|cytolysin|lymphocyte pore forming protein|perforin-1|PRF1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- dG9
- dG9
- Reactivity:
- Human, Bovine
- Human, Bovine
- Host:
- Mouse
- Mouse
- Gene ID:
- 5551
- 5551
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Purified granules from the human lymphoma cell line
- Purified granules from the human lymphoma cell line
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Storage temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT