Order Entry
Canada
ContactUsLinkComponent
Anti-CD29 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HMbeta1-1]
Anti-CD29 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HMbeta1-1]
Supplier:  Biolegend
Certificates
Anti-CD29 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HMbeta1-1]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • CA102205-BL
  • CA102206-BL
  • Antigen symbol:
  • CD29
  • CD29
  • Antigen synonyms:
  • VLA-β chain|GPIIa|b1 integrin|Integrin β1|ITGB1
  • VLA-β chain|GPIIa|b1 integrin|Integrin β1|ITGB1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • HMβ1-1
  • HMβ1-1
  • Reactivity:
  • Rat, Mouse
  • Rat, Mouse
  • Host:
  • Armenian hamster
  • Armenian hamster
  • Gene ID:
  • 16412, 24511
  • 16412, 24511
  • Isotype:
  • IgG
  • IgG
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Purified mouse VLA-4 (α4β1, CD49d/CD29)
  • Purified mouse VLA-4 (α4β1, CD49d/CD29)
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
  • The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
  • Concentration:
  • 0.50 mg/mL
  • 0.50 mg/mL
  • Storage temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together