FITC anti-human CD95 (Fas) [DX2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: CD95
Clonality: Monoclonal
Clone: DX2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Capuchin Monkey;Chimpanzee;Common Marmoset;Cotton-topped Tamarin;Cynomolgus;Pigtailed Macaque;Rhesus;Sooty Mangabey
- Catalog No:
- CA305605-BL
- CA305606-BL
- Antigen symbol:
- CD95
- CD95
- Antigen synonyms:
- FAS|APO-1|TNFRSF6|Fas
- FAS|APO-1|TNFRSF6|Fas
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- DX2
- DX2
- Reactivity:
- Baboon, Capuchin monkey, African green monkey, Cynomolgus monkey, Sooty mangabey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Pigtailed macaque, Chimpanzee, Human
- Baboon, Capuchin monkey, African green monkey, Cynomolgus monkey, Sooty mangabey, Common marmoset, Rhesus monkey, Cotton-topped tamarin, Pigtailed macaque, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 355
- 355
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- CD95 transfected L cells
- CD95 transfected L cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- Storage temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT