Anti-IgG Goat Polyclonal Antibody (DyLight® 680)
Catalogue # CA610-144-003
Fournisseur: Rockland Immunochemical
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:DyLight® 680
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:F(c)
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Mouse IgG F(c) fragment
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA610-144-003
- Supplier No.:610-144-003
Spécifications
À propos de cet article
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope: F(c)
Host: Goat
Isotype:
Reactivity: Mouse