Order Entry
Canada
ContactUsLinkComponent
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 488)
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # CA605-441-013
undefined
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # CA605-441-013
Supplier Number:  605-441-013
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Goat
  • Western blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    Heavy and Light chain
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage temperature:
    –20 °C
  • Concentration:
    1.0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ambient
  • Immunogen:
    Goat IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. no.:
    CA605-441-013
  • Supplier No.:
    605-441-013

Specifications

About this item

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat