Anti-IgG Goat Polyclonal Antibody (DyLight® 405)
Catalog # CA611-146-122
Supplier: Rockland Immunochemical
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Rabbit
- Western blot:Yes
- Size:100 µg
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Rabbit IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA611-146-122
- Supplier No.:611-146-122
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rabbit