Order Entry
Canada
ContactUsLinkComponent
Anti-CD8a Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: RPA-T8]
Anti-CD8a Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: RPA-T8]
Supplier:  Biolegend
Certificates
Anti-CD8a Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: RPA-T8]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • CA301016-BL
  • CA301015-BL
  • Antigen symbol:
  • CD8a
  • CD8a
  • Antigen synonyms:
  • T8|Leu2
  • T8|Leu2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/Cy7®
  • APC (Allophycocyanin)/Cy7®
  • Clone:
  • RPA-T8
  • RPA-T8
  • Reactivity:
  • Baboon, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 925
  • 925
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with APC/Cy7 under optimal conditions. The solution is free of unconjugated APC/Cy7 and unconjugated antibody.
  • The antibody was purified by affinity chromatography, and conjugated with APC/Cy7 under optimal conditions. The solution is free of unconjugated APC/Cy7 and unconjugated antibody.
  • Storage temperature:
  • The CD8a antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The CD8a antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together