Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Anti-TP53BP2 Rabbit Polyclonal Antibody

Anti-TP53BP2 Rabbit Polyclonal Antibody

100 µg
Catalog # CA200-401-A19
Supplier #:  200-401-A19
Anti-TP53BP2 Rabbit Polyclonal Antibody
Catalog # CA200-401-A19
Anti-TP53BP2 Rabbit Polyclonal Antibody
Anti-TP53BP2 Rabbit Polyclonal Antibody

100 µg

Price
$858.14
Log in for availability
Each (100µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Type: Primary
Antigen: TP53BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human

Specifications
  • Taille:
    100 µg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Protein A purified
  • Antibody type:
    Primary
  • Epitope:
    Center
  • Form:
    Liquid (sterile filtered)
  • Isotype:
    IgG
  • Storage temperature:
    –20 °C
  • Host:
    Rabbit
  • Concentration:
    2.4 mg/ml by UV absorbance at 280 nm
  • Cross adsorption:
    No
  • Shipping temperature:
    Dry Ice
  • Antigen name:
    Tumor Protein p53-binding Protein 2
  • Cat. No.:
    CA200-401-A19
  • Reactivity:
    Human
  • Format:
    Antibody fraction
  • Gene ID:
    7159
  • Antigen synonyms:
    P53BP2|ASPP2|PPP1R13A|53BP2|BBP
  • Immunogen:
    This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal sequence of human ASPP2.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    TP53BP2
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.