PharmaScoop Sample Scoops, Bürkle
À propos de cet article
This sample scoop PharmaScoop has been developed in order to meet the high requirements of the pharmaceutical industry, such as those contained in the FDA regulations.
- Stainless steel AISI 316 (1.4404)
- GMP conform
- Highly polished
Like all samplers for the pharmaceutical industry, they are made of stainless steel AISI 316 without edges, crevices or undercuts in which sample residues could collect and contaminate subsequent samples. The highly polished inner and outer surfaces of the scoop therefore meet all the requirements for GMP compliance in production and sampling.
10 Options Available Below
Product Details & Documents
This sample scoop PharmaScoop has been developed in order to meet the high requirements of the pharmaceutical industry, such as those contained in the FDA regulations.
- Stainless steel AISI 316 (1.4404)
- GMP conform
- Highly polished
Like all samplers for the pharmaceutical industry, they are made of stainless steel AISI 316 without edges, crevices or undercuts in which sample residues could collect and contaminate subsequent samples. The highly polished inner and outer surfaces of the scoop therefore meet all the requirements for GMP compliance in production and sampling.
Product Family Options
Informations sur le produit
- DésignationCapacitéLongueurØAvailabilityPrice
- Supplier Number: 5324-0010
- Supplier Number: 5324-0011
- Supplier Number: 5324-0012
- Regular handle500 ml (16.91 oz.)140 mm (5.51")95 mm (3.74")Log in pour la disponibilitéEach$731.00Supplier Number: 5324-0018
- Regular handle750 ml (25.36 oz.)210 mm (8.27")95 mm (3.74")Log in pour la disponibilitéEach$746.09Supplier Number: 5324-0013
- Regular handle1000 ml (33.81 oz.)235 mm (9.25")110 mm (4.33")Log in pour la disponibilitéEach$833.14Supplier Number: 5324-0014
- Regular handle1250 ml (42.27 oz.)195 mm (7.67")125 mm (4.92")Log in pour la disponibilitéEach$827.37Supplier Number: 5324-0015
- Regular handle1700 ml (57.48 oz.)230 mm (9.06")135 mm (5.31")Log in pour la disponibilitéEach$859.39Supplier Number: 5324-0016
- Regular handle2500 ml (84.53 oz.)215 mm (8.46")185 mm (7.28")Log in pour la disponibilitéEach$1,043.48Supplier Number: 5324-0017
- Long handle1000 ml (33.81 oz.)835 mm (32.87")110 mm (4.33")Log in pour la disponibilitéEach$907.56Supplier Number: 5324-0019