Bouin's Solution, Electron Microscopy Sciences | Avantor
Bouin's Solution, Electron Microscopy Sciences
Bouin's Solution, Electron Microscopy Sciences
Certificates
Bouin's Solution, Electron Microscopy Sciences
Bouin's Solution, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Bouin's fixative is used on biopsy specimens of the gastrointestinal tract.
  • Used on biopsy specimens
  • Fixation and staining fluid
Bouin's Solution is used as a fixation and staining fluid. In prefilled leak-proof histo-containers it is ready-to-use. Formulated with Picric Acid saturated aqueous solution 75%, Formalin 25% and Glacial Acetic acid 5% in de-ionized water.
Bouin's fixative is excellent for use on biopsy specimens of the gastrointestinal tract. Tissue from the endocrine system are well fixed and many antibodies react well with tissue fixed in Bouin's. Bouin's solution may be used as a routine fixative or a staining solution, but should not be used as a preservative for tissue. Tissue cannot be held in Bouin's solution for extended periods. The maximum fixation time in this solution should be less than 48 hours and should be transferred to alcohol.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Bouin's Solution
    20 ml
    Discontinued
    Supplier Number: 15990-20
  • Bouin's Solution
    10 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $291.89
    $291.89
    Log in pour la disponibilité
    Each
    Each - $291.89
    $291.89
    Case of 50
       $291.89
    Each
       $291.89
    Supplier Number: 15990-10
  • Bouin's Solution
    30 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $166.47
    $166.47
    Log in pour la disponibilité
    Each
    Each - $166.47
    $166.47
    Case of 20
       $166.47
    Each
       $166.47
    Supplier Number: 15990-30
  • Bouin's Solution
    4 L
    Discontinued
    Supplier Number: 15990-04