KidWind® simpleGEN Classroom Pack
KidWind simpleGEN, class pack of 10, 10
Catalog # CA470165-158
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
: KW-SGENC
KidWind® simpleGEN Classroom Pack
Catalog # CA470165-158
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
The simpleGEN is an easy-to-build AC generator that allows students to explore the basics of electrical generator design.
- Demonstrate core concepts of electricity
- Can be attached to a wind turbine
- Uses alternating current
With the simpleGEN, you can demonstrate Faraday’s law, light LEDs, and perform experiments that explore how coils, magnets, and rotation affect electrical generation. With some quick modifications, you will be able to convert your generator to a simple motor and explore additional variables. You can also take your experiments to the next level by converting your simpleGEN into a wind turbine nacelle. The simpleGEN classroom pack has enough materials to build 10 generators and includes three digital multimeters.
Spécifications
- Désignation:KidWind simpleGEN, class pack of 10
- No. of Lab Groups:10