Utility Jar | Avantor
Utility Jar
Utility Jar
Catalog # 470173-740
Supplier:   Dynalon
Fournisseur #:  600335
Utility Jar
Catalog # 470173-740
Supplier:  Dynalon
Utility Jar
Utility Jar
Utility Jar
Price
$47.75
Log in pour la disponibilité
Each Case of 24
Product Details & Documents
One gallon capacity, high density polyethylene utility jar with metal closure.
  • Wide mouth, natural color
  • Withstands temperatures to 110 °C
Hundreds of uses around the lab.
Spécifications
  • Désignation:
    Utility Jar