Cleanroom Labels, Matte
À propos de cet article
These labels are designed for thermal transfer printing in a cleanroom environment.
- Permenant/removable adhesive
- Made of PP
The special matte finish provides excellent compatibility with cleanroom pens and thermal transfer inks. Available with two different adhesive levels to provide excellent adhesion without leaving residue.
3 Options Available Below
Product Details & Documents
These labels are designed for thermal transfer printing in a cleanroom environment.
- Permenant/removable adhesive
- Made of PP
The special matte finish provides excellent compatibility with cleanroom pens and thermal transfer inks. Available with two different adhesive levels to provide excellent adhesion without leaving residue.
Product Family Options
Informations sur le produit
- DésignationMatièreDimensionsPackagingAvailabilityPrice
- Permenant cleanroom labels, matte finishPP38×38 mm (1.5×1.5")1000 labels/rollLog in pour la disponibilitéRoll of 1,000Roll of 1,000 - $248.64$248.64Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 4000Case of 4000 - $724.14$724.14Prix promotionnel pour les nouveaux clientsRoll of 1,000$248.64Case of 4000$724.14Supplier Number: TX515M
- Permenant cleanroom labels, matte finishPP76×51 mm (3×2")1000 labels/rollDiscontinued-Supplier Number: -
- Removable cleanroom labels, matte finishPP76×51 mm (3×2")1000 labels/rollLog in pour la disponibilitéLog in pour la disponibilitéCase of 4Case of 4 - $799.28$799.28Prix promotionnel pour les nouveaux clientsCase of 4$799.28Supplier Number: TX532M
- Removable cleanroom labels, matte finishPP102×51 mm (4×2")1000 labels/rollLog in pour la disponibilitéRoll of 1,000Roll of 1,000 - $419.90$419.90Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 4000Case of 4000 - $1,187.48$1,187.48Prix promotionnel pour les nouveaux clientsRoll of 1,000$419.90Case of 4000$1,187.48Supplier Number: TX542M