ClipperMop™ Cleanroom Mops | Avantor
ClipperMop™ Cleanroom Mops
Supplier:   Texwipe
ClipperMop™ Cleanroom Mops
Supplier:  Texwipe
Certificates
ClipperMop™ Cleanroom Mops
ClipperMop™ Cleanroom Mops
2 Options Available Below
Product Details & Documents
The mop system may be used with wet, dry or sterile wipers, and is compatible with protocols for ISO Class 3 to 8 cleanrooms, sterile and aseptic areas
  • Cleanroom compatible mopping system
  • Patent-pending thermoplastic mop with clips
  • Quick and easy wiper change
  • Lightweight, easy to handle
  • Less expensive than competitors cleanroom mops
The small size conveniently cleans hard-to-reach areas, walls, ceilings, floors and equipment exteriors. The mop system may be sterilized by gamma irradiation, chemicals, vaporized hydrogen peroxide (VHP) or ethylene oxide. Widely used in pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries.
The patent-pending mophead utilizes non-abrasive polyethylene clips to securely fasten one or multiple wipers to the head. The ClipperMop wiper attachment method does not release particles, fibers and dirt.
Ordering information: The ClipperMop™ cleanroom mop includes the Celcon® thermoplastic mop head with clips, a flexible polyester pad for better wiping efficiency and an adjustable handle.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • ClipperMop™ Cleanroom mop with head assembly and telescoping handle
    18 x 10 cm (7 x 4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $521.90
    $521.90
    Prix promotionnel pour les nouveaux clients
    Each
       $521.90
    Supplier Number: TX7102
  • ClipperMop™ Cleanroom mop with head assembly and telescoping handle
    28 x 10 cm (11 x 4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $543.63
    $543.63
    Prix promotionnel pour les nouveaux clients
    Each
       $543.63
    Supplier Number: TX7103
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ClipperMop™ tether clip replacement kit, 4 polyethylene tether clips and 2 nylon screws per bag
    ClipperMop™ tether clip replacement kit, 4 polyethylene tether clips and 2 nylon screws per bag
    TWTX7102C
    Log in pour la disponibilité
    Case of 20 - $322.04
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ClipperMop™ replacement pads, foam pads (101,6×177,8) and white fasteners for use with TX7103
    ClipperMop™ replacement pads, foam pads (101,6×177,8) and white fasteners for use with TX7103
    TWTX7111B
    Log in pour la disponibilité
    Bag of 10 - $13.58